Learn More
Sino Biological Influenza A H3N2 (A/Victoria/361/2011) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40145V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Victoria/361/2011 (H3N2)) hemagglutinin (AGB08328.1) (Met1-Arg345), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0445