Learn More
Sino Biological Influenza A H3N2 (A/Hong Kong/CUHK31987/2011) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40146V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Hong Kong/CUHK31987/2011(H3N2)) hemagglutinin (AGC13545.1) (Met1-Trp530) , termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0447