Learn More
Sino Biological SARS-CoV Spike/S1 Protein (S1 Subunit, His Tag)
Supplier:  Sino Biological 40150V08B1100
Encompass_Preferred
A DNA sequence encoding the S1 subunit of SARS-CoV (isolate:WH20) spike (AAX16192.1) (Met1-Arg667) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0455