Learn More
Sino Biological SARS-CoV Spike/RBD Protein (RBD, His Tag)
Catalog No. 501620459
Encompass_Preferred
A DNA sequence encoding the receptor binding domain (RBD) of SARS-CoV (isolate:WH20) spike (AAX16192.1) (Arg306-Phe527) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0459 Supplier Sino Biological Supplier No. 40150V08B2100
A DNA sequence encoding the receptor binding domain (RBD) of SARS-CoV (isolate:WH20) spike (AAX16192.1) (Arg306-Phe527) was expressed with a C-terminal polyhistidine tag.