Learn More
Sino Biological SARS-CoV Spike/RBD Protein (RBD, rFc Tag)
Supplier:  Sino Biological 40150V31B2100
Encompass_Preferred
A DNA sequence encoding the receptor binding domain (RBD) of SARS-CoV (isolate:WH20) spike (AAX16192.1) (Arg306-Phe527) was expressed with the Fc region of rabbit IgG at the C-terminus.
Catalog No. 50-162-0463