Learn More
Sino Biological Influenza A H3N2 (A/Victoria/208/2009) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40151V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Victoria/208/2009(H3N2)) hemagglutinin (translated amino acid of EPI189220) (Met1-Arg345), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0469