Learn More
Sino Biological Influenza A H3N2 (A/Guangdong-Luohu/1256/2009) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40152V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Guangdong-Luohu/1256/2009(H3N2)) hemagglutinin (AFM72872.1) (Met1-Arg345), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0475