Learn More
Sino Biological Influenza A H3N2 (A/Babol/36/2005) Hemagglutinin / HA0 Protein
Supplier:  Sino Biological 40153VNAB100
Encompass_Preferred
A DNA sequence encoding the full length of Influenza A virus (A/Babol/36/2005(H3N2)) hemagglutinin (ACN50256.1) (Gln1-Ile550), termed as HA, was expressed.
Catalog No. 50-162-0480