Learn More
Sino Biological Influenza A H3N8 (A/equine/Gansu/7/2008) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40155V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/equine/Gansu/7/2008(H3N8)) hemagglutinin (ACE81938.1) (Met1-Arg344), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0486