Learn More
Sino Biological Influenza B (B/Yamagata/16/1988) Hemagglutinin / HA Protein (ECD, His Tag)
Supplier:  Sino Biological 40157V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza B virus (B/Yamagata/16/1988) hemagglutinin (Met1-Thr546), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0490