Learn More
Sino Biological Influenza B (B/Yamagata/16/1988) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40157V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza B virus (B/Yamagata/16/1988) hemagglutinin (Met1-Arg360), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0492