Learn More
Sino Biological Influenza A H5N1 (A/chicken/VietNam/NCVD-016/2008) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40158V08B2100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/chicken/VietNam/NCVD-016/2008(H5N1)) hemagglutinin (ACO07033.1) (Met1-Gln532) with cleavage site mutated (RRRK →TET, HA1+HA2,uncleaved) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0495