Learn More
MSC Urethane Foam Tape, 3M, 1/16 in Thick, Acrylic, 1 ea
Catalog No. 501113876
Encompass
The 3M Double Coated Urethane Foam Tape 4016 is an off-white, 0.0625 in. (1.6 mm) thick, double-coated, open-cell urethane foam tape offering high shear strength. It is suitable for applications such as mounting interior signage, nameplates, and dispensers.
  • Off-white, 0.0625 in (1.6 mm) open-cell, flexible urethane foam carrier
  • High shear adhesive with high temperature resistance
  • Durable foam ideal for interior mounting applications
  • Ideal for bonding, attaching, and mounting
  • Foam dampens and prevents vibration
  • Conformable foam and adhesive combination provides long-term holding up to 220°F (104°C)
  • Robust, green plaid, densified kraft paper liner for reliable performance in industrial applications
  • Liner helps reduce edge roll and protects from humidity extremes
Catalog No. 50-111-3876 Supplier MSC Supplier No. 65364564
The 3M Double Coated Urethane Foam Tape 4016 is an off-white, 0.0625 in. (1.6 mm) thick, double-coated, open-cell urethane foam tape offering high shear strength. It is suitable for applications such as mounting interior signage, nameplates, and dispensers.
  • Off-white, 0.0625 in (1.6 mm) open-cell, flexible urethane foam carrier
  • High shear adhesive with high temperature resistance
  • Durable foam ideal for interior mounting applications
  • Ideal for bonding, attaching, and mounting
  • Foam dampens and prevents vibration
  • Conformable foam and adhesive combination provides long-term holding up to 220°F (104°C)
  • Robust, green plaid, densified kraft paper liner for reliable performance in industrial applications
  • Liner helps reduce edge roll and protects from humidity extremes