Learn More
Sino Biological Influenza A H5N1 (A/barnswallow/HongKong/D10-1161/2010) Hemagglutinin Protein (HA1+HA2) (His Tag)
Supplier:  Sino Biological 40160V08B1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/barnswallow/HongKong/D10-1161/2010(H5N1)) hemagglutinin (AGC13463.1) (Met1-Gln531) with cleavage site mutated (RRRRK →TET, HA1+HA2, uncleaved) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0501