Learn More
Sino Biological Influenza A H5N8 (A/turkey/Ireland/1378/1983) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40164V08B2100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/turkey/Ireland/1378/1983(H5N8)) hemagglutinin (ABI85117.1) (Met1-Gln527), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0508