Learn More
Sino Biological Influenza A H6N2 (A/duck/Shantou/83/2000) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40166V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/duck/Shantou/83/2000(H6N2)) hemagglutinin (ADG44842.1) (Met1-Gln529), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0516