Learn More
Sino Biological Influenza A H6N5 (A/shearwater/Australia/1/1973) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40167V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/shearwater/Australia/1/1973(H6N5)) hemagglutinin (ABI84516.1) (Met1-Arg344), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0520