Learn More
Sino Biological Influenza A H6N8 (A/mallard/Ohio/217/1998) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40168V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/mallard/Ohio/217/1998(H6N8)) hemagglutinin (ABO52049.1) (Met1-Gln529) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0522