Learn More
Sino Biological Influenza A H6N8 (A/mallard/Ohio/217/1998) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40168V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/mallard/Ohio/217/1998(H6N8)) hemagglutinin (ABO52049.1) (Met1-Arg344), termed as HA1, was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0524