Learn More
Sino Biological Influenza A H7N1 (A/turkey/Italy/4602/99) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40169V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/turkey/Italy/4602/99(H7N1)) hemagglutinin (CAD38286.1) (Met1-Arg339), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0526