Learn More
Sino Biological Influenza A H7N2 (A/ruddy turnstone/New Jersey/563/2006) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40170V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/ruddy turnstone/New Jersey/563/2006(H7N2)) hemagglutinin (ACS68445.1),(Met1-Arg339), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0530