Learn More
Sino Biological Influenza A H7N7 (A/equine/Kentucky/1a/1975) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40171V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/equine/Kentucky/1a/ 1975(H7N7)) hemagglutinin (ACL12085.1) (Met1-Ile534) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0532