Learn More
Sino Biological Influenza A H7N8 (A/mallard/Netherlands/33/2006) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40172V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/mallard/Netherlands/33/2006(H7N8)) hemagglutinin (ACR59554.1) (Met1-Phe535) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0534