Learn More
Sino Biological Influenza A H7N8 (A/mallard/Netherlands/33/2006) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40172V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/mallard/Netherlands/33/2006(H7N8)) hemagglutinin (translated amino acid of EPI182113) (Met1-Arg339), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0536