Learn More
Sino Biological Influenza A H9N2 (A/Hong Kong/35820/2009) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40174V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Hong Kong/35820/ 2009(H9N2)) hemagglutinin (ADC41853.1) (Met1-Lys523) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0539