Learn More
Sino Biological Influenza A H9N5 (A/shorebird/DE/261/2003) Hemagglutinin /HA1 Protein (His Tag)
Supplier:  Sino Biological 40181V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/shorebird/DE/261/2003(H9N5)) hemagglutinin (ABB87950.1) (Met1-Arg338), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0553