Learn More
Sino Biological Influenza A H10N3 (A/mallard/Minnesota/Sg-00194/2007) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40184V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/mallard/Minnesota/Sg-00194/2007(H10N3)) hemagglutinin (ACT84107.1) (Met1-Val525) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0558