Learn More
Sino Biological Influenza A H11N2 (A/thick-billed murre/Newfoundland/031/2007) Hemagglutinin / HA Protein (ECD, His Tag)
Supplier:  Sino Biological 40187V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/thick-billed murre/Newfoundland/031/2007(H11N2)) hemagglutinin (ADD21478.1) (Met1-Lys528), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0562