Learn More
Sino Biological Influenza A H11N2 (A/thick-billed murre/Newfoundland/031/2007) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40187V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/thick-billed murre/Newfoundland/031/2007(H11N2)) (Met1-Arg342), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0564