Learn More
Sino Biological Influenza A H12N3 (A/bar headed goose/Mongolia/143/2005) Hemagglutinin / HA1 Protein (His Tag)
Supplier:  Sino Biological 40189V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/bar headed goose/Mongolia/143/2005(H12N3)) hemagglutinin (ACV86810.1), (Met1-Arg342) termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0568