Learn More
Sino Biological Influenza A H13N6 (A/black-headed gull/Sweden/1/1999) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40190V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/black-headed gull/Sweden/1/1999(H13N6)) hemagglutinin (AAV91213.1) (Met1-Arg342), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0570