Learn More
Sino Biological Influenza B (B/Massachusetts/03/2010) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40191V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza B virus (B/Massachusetts/03/2010) hemagglutinin ( AGB07680.1) (Met1-Thr548), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0574