Learn More
Sino Biological Influenza B (B/Massachusetts/03/2010) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40191V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza B virus (B/Massachusetts/03/2010) hemagglutinin (AGI64446.1) (Met1-Arg362), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0576