Learn More
Sino Biological Influenza B (B/Massachusetts/03/2010) Hemagglutinin / HA0 Protein
Supplier:  Sino Biological 40191VNAB100
Encompass_Preferred
A DNA sequence encoding the full length of Influenza B virus (B/Massachusetts/03/2010) hemagglutinin ( AGB07680.1) (Met1-Leu585), termed as HA, was expressed.
Catalog No. 50-162-0579