Learn More
Sino Biological Influenza A H15N2 (A/Australian shelduck/Western Australia/1756/1983) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40193V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Australian shelduck/Western Australia/1756/1983(H15N2)) hemagglutinin (ABB90704.1) (Met1-Val534) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0585