Learn More
Sino Biological Influenza A H1N1 (A/USSR/90/1977) Neuraminidase / NA (His Tag)
Supplier:  Sino Biological 40197V07H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/USSR/90/1977 (H1N1)) neuraminidase (P03469.2) (His36-Lys470) was expressed with a N-terminal polyhistidine tag.
Catalog No. 50-162-0590