Learn More
Sino Biological Influenza A H3N2 (A/Aichi/2/1968) Neuraminidase / NA (His Tag)
Catalog No. 501620595
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Aichi/2/1968(H3N2)) neuraminidase (Q75VQ4.1) (His36-Ile469) was expressed with a N-terminal polyhistidine tag.
Catalog No. 50-162-0595 Supplier Sino Biological Supplier No. 40199V07H100
A DNA sequence encoding the Influenza A virus (A/Aichi/2/1968(H3N2)) neuraminidase (Q75VQ4.1) (His36-Ile469) was expressed with a N-terminal polyhistidine tag.