Learn More
Sino Biological Influenza A H7N7 (A/Netherlands/219/2003) Neuraminidase / NA Protein (His Tag)
Supplier:  Sino Biological 40202V07H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Netherlands/219/2003(H7N7)) neuraminidase (AAR11367.1) (His36-Ser471) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-0598