Learn More
Sino Biological Influenza A H1N1 (A/California/07/2009) Nucleoprotein / NP Protein (His Tag)
Supplier:  Sino Biological 40205V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/California/07/2009(H1N1)) nucleoprotein (ACS94534.1) (Met1-Ser498) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-0607