Learn More
Aviva Systems Biology Corp MARCH2 Antibody - C-terminal region (ARP43220_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43220_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MARCH2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MARCH2. Alternate names: HSPC240, MARCH-II, RNF172ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58463