Learn More
Aviva Systems Biology Corp MARCH5 Antibody - N-terminal region (ARP43232_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43232_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MARCH5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MARCH5. Alternate names: FLJ20445, MARCH-V, MITOL, RNF153ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58466