Learn More
Sino Biological HIV-1 gp140 Protein (group M, subtype A, strain 92UG037.1) (Fc Tag)
Supplier:  Sino Biological 40242V02H100
Encompass_Preferred
A DNA sequence encoding the HIV-1 envelope glycoprotein gp160 (AAC97548.1) extracellular domain (Leu33-Lys659), termed as gp140, was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-0630