Learn More
Sino Biological HIV-1 p24 Protein (group M, subtype B, strain HXB2) (His Tag)
Supplier:  Sino Biological 40243V08E100
Encompass_Preferred
A DNA sequence encoding the HIV1 Gag-P24 (subtype B) (AAB50258.1) (Pro133-Leu363) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0634