Learn More
Aviva Systems Biology Corp MARCH7 Antibody - middle region (ARP43307_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43307_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MARCH7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MARCH7. Alternate names: AXO, AXOT, DKFZP586F1122, MARCH-VII, RNF177ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58490