Learn More
Sino Biological HIV-p51 / RT-p51 (group M, subtype B (isolate HXB2) Gag-Pol polyprotein Protein (His Tag)
Supplier:  Sino Biological 40244V07E100
Encompass_Preferred
A DNA sequence encoding the p51 (Reverse Transcriptase)of human immunodeficiency virus 1 gag-pol (NP_057849.4) (Pro588-Phe1027) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-0638