Learn More
Sino Biological HIV-p66 / RT-p66 (group M, subtype B (isolate HXB2) Gag-Pol polyprotein Protein (His Tag)
Supplier:  Sino Biological 40244V07E1100
Encompass_Preferred
A DNA sequence encoding the p66 (Reverse Transcriptase + RNase H) of human immunodeficiency virus 1 gag-pol (NP_057849.4) (Pro588-Leu1147) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-0639