Learn More
Aviva Systems Biology Corp MARCH8 Antibody - N-terminal region (ARP42958_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42958_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MARCH8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MARCH8. Alternate names: MARCH-VIII, MIR, RNF178, c-MIRApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rat
Catalog No. 50-014-58372