Learn More
Aviva Systems Biology Corp MARCH8 Antibody - middle region (ARP43401_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43401_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MARCH8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MARCH8. Alternate names: MARCH-VIII, MIR, RNF178, c-MIRApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58521