Learn More
Sino Biological Human immunodeficiency virus 1 (HIV-1) (group M, subtype C, strain 92BR025) gp140 Protein (Fc Tag)
Supplier:  Sino Biological 40251V02H100
Encompass_Preferred
A DNA sequence encoding the Human immunodeficiency virus 1 (HIV1) (group M, subtype C, strain 92BR025) GP140 (AAB61124.1) (Asn32-Ile677) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-0650