Learn More
Sino Biological HIV-1 p24 Protein (group O, strain BCF06) (His Tag)
Supplier:  Sino Biological 40256V08E100
Encompass_Preferred
A DNA sequence encoding the HIV1 Gag-P24 (group O) (BAH97658.1) (Pro131-Leu362) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-0666